CD11b Monoclonal Antibody (ICRF44), APC-eFluor™ 780, eBioscience™ - FAQs
CD11b Monoclonal Antibody (ICRF44), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD11b Monoclonal Antibody (ICRF44), APC-eFluor™ 780, eBioscience™ - FAQs (47-0118-41, 47-0118-42)