CD11c Monoclonal Antibody (BU15), APC-eFluor™ 780, eBioscience™ - FAQs
CD11c Monoclonal Antibody (BU15), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (BU15), APC-eFluor™ 780, eBioscience™ - FAQs (47-0128-42, 47-0128-41)