CD14 Monoclonal Antibody (61D3), APC-eFluor™ 780, eBioscience™ - FAQs
CD14 Monoclonal Antibody (61D3), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD14 Monoclonal Antibody (61D3), APC-eFluor™ 780, eBioscience™ - FAQs (47-0149-42, 47-0149-41)