CD19 Monoclonal Antibody (SJ25C1), APC-eFluor™ 780, eBioscience™ - FAQs
CD19 Monoclonal Antibody (SJ25C1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (SJ25C1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0198-42, 47-0198-41)