CD19 Monoclonal Antibody (HIB19), APC-eFluor™ 780, eBioscience™ - FAQs
CD19 Monoclonal Antibody (HIB19), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (HIB19), APC-eFluor™ 780, eBioscience™ - FAQs (47-0199-41, 47-0199-42)