CD20 Monoclonal Antibody (2H7), APC-eFluor™ 780, eBioscience™ - FAQs
CD20 Monoclonal Antibody (2H7), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD20 Monoclonal Antibody (2H7), APC-eFluor™ 780, eBioscience™ - FAQs (47-0209-41, 47-0209-42)