CD23 Monoclonal Antibody (EBVCS2), APC-eFluor™ 780, eBioscience™ - FAQs
CD23 Monoclonal Antibody (EBVCS2), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD23 Monoclonal Antibody (EBVCS2), APC-eFluor™ 780, eBioscience™ - FAQs (47-0238-42)