CD24 Monoclonal Antibody (M1/69), APC-eFluor™ 780, eBioscience™ - FAQs
CD24 Monoclonal Antibody (M1/69), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD24 Monoclonal Antibody (M1/69), APC-eFluor™ 780, eBioscience™ - FAQs (47-0242-82, 47-0242-80)