CD25 Monoclonal Antibody (PC61.5), APC-eFluor™ 780, eBioscience™ - FAQs
CD25 Monoclonal Antibody (PC61.5), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (PC61.5), APC-eFluor™ 780, eBioscience™ - FAQs (47-0251-82, 47-0251-80)