CD25 Monoclonal Antibody (CD25-4E3), APC-eFluor™ 780, eBioscience™ - FAQs
CD25 Monoclonal Antibody (CD25-4E3), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (CD25-4E3), APC-eFluor™ 780, eBioscience™ - FAQs (47-0257-42, 47-0257-41)