CD25 Monoclonal Antibody (BC96), APC-eFluor™ 780, eBioscience™ - FAQs
CD25 Monoclonal Antibody (BC96), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (BC96), APC-eFluor™ 780, eBioscience™ - FAQs (47-0259-41, 47-0259-42)