CD27 Monoclonal Antibody (LG.7F9), APC-eFluor™ 780, eBioscience™ - FAQs
CD27 Monoclonal Antibody (LG.7F9), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD27 Monoclonal Antibody (LG.7F9), APC-eFluor™ 780, eBioscience™ - FAQs (47-0271-80, 47-0271-82)