CD27 Monoclonal Antibody (O323), APC-eFluor™ 780, eBioscience™ - FAQs
CD27 Monoclonal Antibody (O323), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD27 Monoclonal Antibody (O323), APC-eFluor™ 780, eBioscience™ - FAQs (47-0279-41, 47-0279-42)