CD28 Monoclonal Antibody (CD28.2), APC-eFluor™ 780, eBioscience™ - FAQs
CD28 Monoclonal Antibody (CD28.2), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD28 Monoclonal Antibody (CD28.2), APC-eFluor™ 780, eBioscience™ - FAQs (47-0289-41, 47-0289-42)