CD34 Monoclonal Antibody (4H11), APC-eFluor™ 780, eBioscience™ - FAQs
CD34 Monoclonal Antibody (4H11), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD34 Monoclonal Antibody (4H11), APC-eFluor™ 780, eBioscience™ - FAQs (47-0349-42, 47-0349-41)