CD38 Monoclonal Antibody (HIT2), APC-eFluor™ 780, eBioscience™ - FAQs
CD38 Monoclonal Antibody (HIT2), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD38 Monoclonal Antibody (HIT2), APC-eFluor™ 780, eBioscience™ - FAQs (47-0389-42, 47-0389-41)