CD39 Monoclonal Antibody (24DMS1), APC-eFluor™ 780, eBioscience™ - FAQs
CD39 Monoclonal Antibody (24DMS1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD39 Monoclonal Antibody (24DMS1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0391-80, 47-0391-82)