CD41a Monoclonal Antibody (eBioMWReg30 (MWReg30)), APC-eFluor™ 780, eBioscience™ - FAQs
CD41a Monoclonal Antibody (eBioMWReg30 (MWReg30)), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD41a Monoclonal Antibody (eBioMWReg30 (MWReg30)), APC-eFluor™ 780, eBioscience™ - FAQs (47-0411-80, 47-0411-82)