CD45 Monoclonal Antibody (30-F11), APC-eFluor™ 780, eBioscience™ - FAQs
CD45 Monoclonal Antibody (30-F11), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (30-F11), APC-eFluor™ 780, eBioscience™ - FAQs (47-0451-82, 47-0451-80)