CD45.2 Monoclonal Antibody (104), APC-eFluor™ 780, eBioscience™ - FAQs
CD45.2 Monoclonal Antibody (104), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD45.2 Monoclonal Antibody (104), APC-eFluor™ 780, eBioscience™ - FAQs (47-0454-82, 47-0454-80)