CD45RA Monoclonal Antibody (HI100), APC-eFluor™ 780, eBioscience™ - FAQs
CD45RA Monoclonal Antibody (HI100), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD45RA Monoclonal Antibody (HI100), APC-eFluor™ 780, eBioscience™ - FAQs (47-0458-42, 47-0458-41)