CD45 Monoclonal Antibody (HI30), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (HI30), APC-eFluor™ 780, eBioscience™ - FAQs (47-0459-41, 47-0459-42)