CD45 Monoclonal Antibody (OX1), APC-eFluor™ 780, eBioscience™ - FAQs
CD45 Monoclonal Antibody (OX1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (OX1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0461-82, 47-0461-80)