CD48 Monoclonal Antibody (HM48-1), APC-eFluor™ 780, eBioscience™ - FAQs
CD48 Monoclonal Antibody (HM48-1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD48 Monoclonal Antibody (HM48-1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0481-82, 47-0481-80)