CD64 Monoclonal Antibody (10.1), APC-eFluor™ 780, eBioscience™ - FAQs
CD64 Monoclonal Antibody (10.1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD64 Monoclonal Antibody (10.1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0649-41)