CD69 Monoclonal Antibody (H1.2F3), APC-eFluor™ 780, eBioscience™ - FAQs
CD69 Monoclonal Antibody (H1.2F3), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD69 Monoclonal Antibody (H1.2F3), APC-eFluor™ 780, eBioscience™ - FAQs (47-0691-82, 47-0691-80)