CD69 Monoclonal Antibody (FN50), APC-eFluor™ 780, eBioscience™ - FAQs
CD69 Monoclonal Antibody (FN50), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD69 Monoclonal Antibody (FN50), APC-eFluor™ 780, eBioscience™ - FAQs (47-0699-41, 47-0699-42)