CD79a Monoclonal Antibody (HM47), APC-eFluor™ 780, eBioscience™ - FAQs
CD79a Monoclonal Antibody (HM47), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD79a Monoclonal Antibody (HM47), APC-eFluor™ 780, eBioscience™ - FAQs (47-0792-41, 47-0792-42)