CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), APC-eFluor™ 780, eBioscience™ - FAQs
CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0902-82)