CD115 (c-fms) Monoclonal Antibody (AFS98), APC-eFluor™ 780, eBioscience™ - FAQs
CD115 (c-fms) Monoclonal Antibody (AFS98), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD115 (c-fms) Monoclonal Antibody (AFS98), APC-eFluor™ 780, eBioscience™ - FAQs (47-1152-82, 47-1152-80)