CD117 (c-Kit) Monoclonal Antibody (2B8), APC-eFluor™ 780, eBioscience™ - FAQs
CD117 (c-Kit) Monoclonal Antibody (2B8), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD117 (c-Kit) Monoclonal Antibody (2B8), APC-eFluor™ 780, eBioscience™ - FAQs (47-1171-80, 47-1171-82)