CD123 Monoclonal Antibody (6H6), APC-eFluor™ 780, eBioscience™ - FAQs
CD123 Monoclonal Antibody (6H6), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD123 Monoclonal Antibody (6H6), APC-eFluor™ 780, eBioscience™ - FAQs (47-1239-41, 47-1239-42)