CD127 Monoclonal Antibody (A7R34), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD127 Monoclonal Antibody (A7R34), APC-eFluor™ 780, eBioscience™ - FAQs (47-1271-80, 47-1271-82)