CD127 Monoclonal Antibody (eBioRDR5), APC-eFluor™ 780, eBioscience™ - FAQs
CD127 Monoclonal Antibody (eBioRDR5), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD127 Monoclonal Antibody (eBioRDR5), APC-eFluor™ 780, eBioscience™ - FAQs (47-1278-41, 47-1278-42)