CD197 (CCR7) Monoclonal Antibody (4B12), APC-eFluor™ 780, eBioscience™ - FAQs
CD197 (CCR7) Monoclonal Antibody (4B12), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD197 (CCR7) Monoclonal Antibody (4B12), APC-eFluor™ 780, eBioscience™ - FAQs (47-1971-82, 47-1971-80)