CD197 (CCR7) Monoclonal Antibody (3D12), APC-eFluor™ 780, eBioscience™ - FAQs
CD197 (CCR7) Monoclonal Antibody (3D12), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD197 (CCR7) Monoclonal Antibody (3D12), APC-eFluor™ 780, eBioscience™ - FAQs (47-1979-41, 47-1979-42)