CD304 (Neuropilin-1) Monoclonal Antibody (TNKUSOHA), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD304 (Neuropilin-1) Monoclonal Antibody (TNKUSOHA), APC-eFluor™ 780, eBioscience™ - FAQs (47-3049-42, 47-3049-41)