CD370 (Clec9A) Monoclonal Antibody (9A11), APC-eFluor™ 780, eBioscience™ - FAQs
CD370 (Clec9A) Monoclonal Antibody (9A11), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD370 (Clec9A) Monoclonal Antibody (9A11), APC-eFluor™ 780, eBioscience™ - FAQs (47-3709-41, 47-3709-42)