Rat IgG2b kappa Isotype Control (eB149/10H5), APC-eFluor™ 780, eBioscience™ - FAQs
Rat IgG2b kappa Isotype Control (eB149/10H5), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Rat IgG2b kappa Isotype Control (eB149/10H5), APC-eFluor™ 780, eBioscience™ - FAQs (47-4031-82, 47-4031-80)