Rat IgG1 kappa Isotype Control (eBRG1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Rat IgG1 kappa Isotype Control (eBRG1), APC-eFluor™ 780, eBioscience™ - FAQs (47-4301-80)