eBioscience™ Streptavidin APC-eFluor™ 780 Conjugate - FAQs
eBioscience™ Streptavidin APC-eFluor™ 780 Conjugate - FAQs
View additional product information for eBioscience™ Streptavidin APC-eFluor™ 780 Conjugate - FAQs (47-4317-82)