Rat IgG2a kappa Isotype Control (eBR2a), APC-eFluor™ 780, eBioscience™ - FAQs
Rat IgG2a kappa Isotype Control (eBR2a), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), APC-eFluor™ 780, eBioscience™ - FAQs (47-4321-82, 47-4321-80)