Mouse IgG2b kappa Isotype Control (eBMG2b), APC-eFluor™ 780, eBioscience™ - FAQs
Mouse IgG2b kappa Isotype Control (eBMG2b), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Mouse IgG2b kappa Isotype Control (eBMG2b), APC-eFluor™ 780, eBioscience™ - FAQs (47-4732-80)