F4/80 Monoclonal Antibody (BM8), APC-eFluor™ 780, eBioscience™ - FAQs
F4/80 Monoclonal Antibody (BM8), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for F4/80 Monoclonal Antibody (BM8), APC-eFluor™ 780, eBioscience™ - FAQs (47-4801-80, 47-4801-82)