Armenian Hamster IgG Isotype Control (eBio299Arm), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Armenian Hamster IgG Isotype Control (eBio299Arm), APC-eFluor™ 780, eBioscience™ - FAQs (47-4888-80)