Ki-67 Monoclonal Antibody (SolA15), APC-eFluor™ 780, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (SolA15), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (SolA15), APC-eFluor™ 780, eBioscience™ - FAQs (47-5698-80, 47-5698-82)
This product does not have any FAQs available.