Mouse IgM Monoclonal Antibody (II/41), APC-eFluor™ 780, eBioscience™ - FAQs
Mouse IgM Monoclonal Antibody (II/41), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Mouse IgM Monoclonal Antibody (II/41), APC-eFluor™ 780, eBioscience™ - FAQs (47-5790-80, 47-5790-82)