TCR V alpha 2 Monoclonal Antibody (B20.1), APC-eFluor™ 780, eBioscience™ - FAQs
TCR V alpha 2 Monoclonal Antibody (B20.1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for TCR V alpha 2 Monoclonal Antibody (B20.1), APC-eFluor™ 780, eBioscience™ - FAQs (47-5812-80)