KLRG1 Monoclonal Antibody (2F1), APC-eFluor™ 780, eBioscience™ - FAQs
KLRG1 Monoclonal Antibody (2F1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for KLRG1 Monoclonal Antibody (2F1), APC-eFluor™ 780, eBioscience™ - FAQs (47-5893-80, 47-5893-82)