FceR1 alpha Monoclonal Antibody (MAR-1), APC-eFluor™ 780, eBioscience™ - FAQs
FceR1 alpha Monoclonal Antibody (MAR-1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for FceR1 alpha Monoclonal Antibody (MAR-1), APC-eFluor™ 780, eBioscience™ - FAQs (47-5898-82, 47-5898-80)